
3-Hydroxy-o-toluic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:603-80-5
:
Formula:C₈H₈O₃
MDL Number:MFCD00671541
:152.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:143.0 to 147.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together