Order Entry
Belgium
ContactUsLinkComponent
N-Ethyldiisopropylamine ∼10% in DMF for detection of primary amines
Flammable
Corrosive
Irritant
Health
N-Ethyldiisopropylamine ∼10% in DMF for detection of primary amines
Catalog # TCIAD2937-10ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
N-Ethyldiisopropylamine ∼10% in DMF for detection of primary amines
Catalog # TCIAD2937-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Nonaqueous Titration):
    9.0 to 12.0 w/w%

Specification Test Results

About this item

 :
 :129.25 g/mol
Storage Temperature:Ambient
EINECS:230-392-0
UN:2733
ADR:3,III
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868