Order Entry
Belgium
ContactUsLinkComponent
Fluoride standard solution, 0.2 mg/L F- in Water (from NaF) according to DIN 38405-4:1985, DEV D4
Fluoride standard solution, 0.2 mg/L F- in Water (from NaF) according to DIN 38405-4:1985, DEV D4
Catalog # KRAF20639.2000
Supplier:  Bernd Kraft
CAS Number:  
Fluoride standard solution, 0.2 mg/L F- in Water (from NaF) according to DIN 38405-4:1985, DEV D4
Catalog # KRAF20639.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    0,2 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Fluoride standard solution
  • Contents:
    NaF in H₂O

Specifications

About this item