Selenium standard solution, 100 mg/L Se in dil. nitric acid standard for ICP-MS
Catalog # ACSDM352.2NP.L1
Supplier: CPACHEM
CAS Number:
Specifications
- Concentration:100 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Selenium
- Symbol:Se