Order Entry
Belgium
ContactUsLinkComponent
Cobalt standard solution, 100 mg/L Co in dil. nitric acid standard for ICP-MS
Cobalt standard solution, 100 mg/L Co in dil. nitric acid standard for ICP-MS
Catalog # ACSDM312.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Cobalt standard solution, 100 mg/L Co in dil. nitric acid standard for ICP-MS
Catalog # ACSDM312.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item