Order Entry
Belgium
ContactUsLinkComponent
Antimony standard solution, 10000 mg/L Sb in 10% nitric acid with 2% hydrofluoric acid standard for ICP
Antimony standard solution, 10000 mg/L Sb in 10% nitric acid with 2% hydrofluoric acid standard for ICP
Catalog # ACSDC150.10N2FP.L5
Supplier:  CPACHEM
CAS Number:  
Antimony standard solution, 10000 mg/L Sb in 10% nitric acid with 2% hydrofluoric acid standard for ICP
Catalog # ACSDC150.10N2FP.L5
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    10% HNO₃+2% HF
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Antimony
  • Symbol:
    Sb

Specifications

About this item