Order Entry
Belgium
ContactUsLinkComponent
 
Antimony standard solution, 10 mg/L Sb in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP-MS
Catalog # 89190.180
Supplier:  CPACHEM
CAS Number:  
Antimony standard solution, 10 mg/L Sb in dil. nitric acid with hydrofluoric acid (max. 1%) standard for ICP-MS
Catalog # 89190.180
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Antimony
  • Symbol:
    Sb

Specifications

About this item