Order Entry
Belgium
ContactUsLinkComponent
Palladium standard solution, 100 mg/L Pd in 2% nitric acid standard for ICP-MS
Palladium standard solution, 100 mg/L Pd in 2% nitric acid standard for ICP-MS
Catalog # ACSDM342.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Palladium standard solution, 100 mg/L Pd in 2% nitric acid standard for ICP-MS
Catalog # ACSDM342.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Palladium
  • Symbol:
    Pd

Specifications

About this item