Order Entry
Belgium
ContactUsLinkComponent
Aluminium standard solution, 100 mg/L Al in dil. nitric acid standard for ICP-MS
Aluminium standard solution, 100 mg/L Al in dil. nitric acid standard for ICP-MS
Catalog # ACSDM302.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Aluminium standard solution, 100 mg/L Al in dil. nitric acid standard for ICP-MS
Catalog # ACSDM302.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item