Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
 
Tetrabutylammonium hydroxide 10% in methanol for non-aqueous titration
 :  TCI
 
undefined
 
 
 
 
Tetrabutylammonium hydroxide 10% in methanol for non-aqueous titration
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    10.0 to 11.0 w/w%
  • Water:
    max. 2.0 %

 

Frequently Bought Together