Order Entry
Belgium
ContactUsLinkComponent
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC, titration analysis)
Flammable
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Flammable
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together