Order Entry
Belgium
ContactUsLinkComponent
2,1,3-Benzothiadiazole-4-sulphonyl chloride
2,1,3-Benzothiadiazole-4-sulphonyl chloride
Catalog # MAYBKM06358.0003
Supplier:  Thermo Scientific
CAS Number:  
2,1,3-Benzothiadiazole-4-sulphonyl chloride
Catalog # MAYBKM06358.0003
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

About this item

 :
 :234.68 g/mol
UN:3261
ADR:8,III
CAS Number:73713-79-8
Formula:C₆H₃ClN₂O₂S₂
MDL Number:MFCD00068049