2-Acrylamido-2-methylpropanesulphonic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15214-89-8
:
Formula:C₇H₁₃NO₄S
MDL Number:MFCD00007522
:207.25 g/mol
EINECS:239-268-0
UN:3261
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Bromination Method):min. 98.0 %
Specification Test Results
Frequently Bought Together