Order Entry
Belgium
ContactUsLinkComponent
1,4-Bis(methylamino)-9,10-anthraquinone
1,4-Bis(methylamino)-9,10-anthraquinone
Catalog # TCIAD3280-25G
Supplier:  TCI
CAS Number:  
undefined
1,4-Bis(methylamino)-9,10-anthraquinone
Catalog # TCIAD3280-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Dark purple powder to crystal
  • Lambda max.:
    641.0 to 645.0 nm (EtOH)
  • Lambda max.2:
    593.0 to 597.0 nm (EtOH)
  • Absorbance (E1%1cm):
    min. 600 (EtOH, 641.0 to 645.0 nm)
  • Absorbance (E1%1cm)2:
    min. 510 (EtOH, 593.0 to 597.0 nm)

Specification Test Results

About this item

Synonyms: 1,4-Bis(methylamino)anthraquinone, Disperse Blue 14, 1,4-Bis(methylamino)anthracene-9,10-dione
 :
 :266.29 g/mol
EINECS:219-602-1
 :GHS07
 :GHS08
CAS Number:2475-44-7
Formula:C₁₆H₁₄N₂O₂
MDL Number:MFCD00001198
Melting Pt:222°C