2-Amino-4-(2-naphthyl)thiazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-(Naphthalen-2-yl)thiazol-2-amine
CAS Number:21331-43-1
:
Formula:C₁₃H₁₀N₂S
MDL Number:MFCD00046452
Molecular Weight:226.30 g/mol
Melting Pt:153…156 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together