Order Entry
Belgium
ContactUsLinkComponent
 
Fluorescein diacetate 97%, pure
 
undefined
Fluorescein diacetate 97%, pure

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    200 to 205 °C
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥96.0 %

 

 

Frequently Bought Together