TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Catalog # MOLEM10255546
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: TBTU
:
:321.1 g/mol
CAS Number:125700-67-6
Formula:C₁₁H₁₆BF₄N₅O
MDL Number:MFCD00077413