
Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilised, powder -100+325 Mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-23-4
:
Formula:ZrO₂
MDL Number:MFCD00011310
:123.22 g/mol
Merck Index:14,10180
Boiling Pt:∼ 5000 °C
Melting Pt:∼ 2700 °C
Density:5.75 g/cm³ (20 °C)
:Stabilised with calcia (CaO) 4%
EINECS:215-227-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together