Fmoc-D-phenylalanine ≥98.0% (par HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:86123-10-6
:
Formula:C₂₄H₂₁NO₄
MDL Number:MFCD00062955
:387.4 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 96.0 %
- Melting point:182.0 to 187.0 deg-C
- Specific rotation [a]20/D:+37.0 to +41.0 deg (C=1, DMF)
Specification Test Results
Frequently Bought Together