Lactate standard solution, 1000 mg/L CH3CH(OH)COO- in Water standard for ion chromatography
: CPACHEM
Lactate standard solution, 1000 mg/L CH3CH(OH)COO- in Water standard for ion chromatography
: CPACHEM
Frequently Bought Together
pH buffers, DuraCal
: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
pH buffer solutions, NIST traceable, Orion™
: Thermo Orion
Starch Potassium Iodide Paper Reag. Ph. Eur. 1085106
: VWR Chemicals
1,2,4-TRIAZOLE 1 * 100 mg
: Merck
SILAC HI3 E. COLI STANDARD 1 * 1 KIT
: WATERS
Buffer solutions in sachets
: Mettler - Toledo
Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
: Thermo Fisher Scientific
VWR®, Turbidity standards, formazin
: VWR Chemicals