ContactUsLinkComponent
HPLC columns, sugar analysis, NUCLEOGEL® SUGAR 810
HPLC columns, sugar analysis, NUCLEOGEL® SUGAR 810
Supplier:  MACHEREY-NAGEL
Certificates
undefined
HPLC columns, sugar analysis, NUCLEOGEL® SUGAR 810
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA719575
  • MANA719574
  • MANA719571
  • MANA719570
  • Description:
  • SUGAR 810 H
  • SUGAR 810 H
  • SUGAR 810 Ca
  • SUGAR 810 Ca
  • Max. pressure:
  • 70 bar
  • 70 bar
  • 70 bar
  • 70 bar
  • Temperature limit:
  • 50 - 90 °C
  • 50 - 90 °C
  • 50 - 90 °C
  • 50 - 90 °C
  • USP code:
  • L17
  • L17
  • L19
  • L19