SPE columns, CHROMABOND® OH (Diol)
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® OH is a diol modified silica phase for SPE.
- Polar phase
- Distinct hydrogen bonds
- Interaction with polar compounds
Recommended for polar substances like antibiotics and prostaglandins.
Specifications
- Catalog No:
- MANA730053
- MANA730418
- Carbon load:
- 5,5%
- 5,5%
- pH range:
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g