Disposable earplugs, Pura-Fit®
Supplier: Moldex
Certificates
About this item
These plugs made of soft PU foam provide protection against higher noise levels.
- Tapered form makes it easy to insert the plugs in the ear
- Outer surface is smooth, so it doesn't irritate the canal
- Bright colour for improved visibility
SNR: 36 dB
Certifications: EN 352-2:2002
Packaging: As pairs or in the hygienic dispenser system MoldexStation.
Specifications
- Catalog No:
- MOLD7700
- MOLD6900
- MOLD7750
- Environmentally Preferable:
- Safer Human and Environmental Health