Order Entry
Belgium
ContactUsLinkComponent
Decontamination solution for autoclaves, Autoclave-ExitusPlus™
Decontamination solution for autoclaves, Autoclave-ExitusPlus™
N° de catalogue A7600.1000
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Decontamination solution for autoclaves, Autoclave-ExitusPlus™
N° de catalogue A7600.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    6 L
  • Description:
    Autoclave-ExitusPlus™, 6× 1 l
  • Storage temperature:
    Ambient

Specifications

About this item

Autoclaving is badly suited for the sustained removal of nucleic acids. DNA molecules in recombinant microorganisms are merely fragmented in the autoclave and subsequently released with the vapours when opening. By adding Autoclave-ExitusPlus™ to residual cultures or buffer solutions, the nucleic acids are effectively destroyed during autoclaving.