ContactUsLinkComponent
Accurate digital refactometers, RX-i
Accurate digital refactometers, RX-i
Supplier:  Atago
Certificates
undefined
Accurate digital refactometers, RX-i
Supplier:  Atago
Specifications
  • Catalog No:
  • ATAG3276
  • ATAG3278
  • ATAG3275
  • ATAG3279
  • Display:
  • Colour touch screen
  • Colour touch screen
  • Colour touch screen
  • Colour touch screen
  • Prism:
  • Artificial sapphire
  • Artificial sapphire
  • Artificial sapphire
  • Artificial sapphire
  • Temperature measurement:
  • 5...75 °C
  • 5...75 °C
  • 5...75 °C
  • 5...75 °C
  • Temperature stability:
  • ±0,05 °C
  • ±0,05 °C
  • ±0,05 °C
  • ±0,05 °C
  • W×D×H:
  • 370×260×140 mm
  • 370×260×140 mm
  • 370×260×140 mm
  • 370×260×140 mm