Orders LinkContactUsLinkComponent
Dropping bottles, LDPE
Dropping bottles, LDPE
Supplier:   VITLAB
Dropping bottles, LDPE
Supplier:  VITLAB
Certificates
Dropping bottles, LDPE
Dropping bottles, LDPE
5 Options Available Below
Product Details & Documents
LDPE, translucent.
  • With dropper insert and HDPE screw cap
  • Used for food safe products
Transparent narrow mouth bottles with extra-long, fine dropping tip for accurate dispensing.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 50 ml
    129 mm
    37 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 106,00 €
    106,00 €
    1 * 24 ST
       106,00 €
  • 100 ml
    155 mm
    43 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 108,00 €
    108,00 €
    1 * 24 ST
       108,00 €
  • 250 ml
    183 mm
    65 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 88,70 €
    88,70 €
    1 * 12 ST
       88,70 €
  • 500 ml
    228 mm
    74 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 93,80 €
    93,80 €
    1 * 12 ST
       93,80 €
  • 1000 ml
    269 mm
    92 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 107,00 €
    107,00 €
    1 * 12 ST
       107,00 €