Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
Supplier:   COWIE
Stoppers, self realasing, PTFE
Supplier:  COWIE
Certificates
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
9 Options Available Below
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Product Family Options
Product Information
  • NS
    Height
    Availability
    Price
  • 19/26
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,10 €
    20,10 €
    1 * 1 ST
       20,10 €
  • 24/29
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,00 €
    27,00 €
    1 * 1 ST
       27,00 €
  • 29/32
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,40 €
    32,40 €
    1 * 1 ST
       32,40 €
  • 34/35
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,10 €
    34,10 €
    1 * 1 ST
       34,10 €
  • 45/40
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,20 €
    60,20 €
    1 * 1 ST
       60,20 €
  • 45/50
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,60 €
    65,60 €
    1 * 1 ST
       65,60 €
  • 34/35
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,30 €
    37,30 €
    1 * 1 ST
       37,30 €
  • 19/38
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,00 €
    23,00 €
    1 * 1 ST
       23,00 €
  • 24/40
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,50 €
    29,50 €
    1 * 1 ST
       29,50 €