Volumetric flasks, class B, PP
Supplier: VITLAB
Certificates
About this item
PP, highly transparent, with PP screw cap.
- With ring mark individually calibrated to ‘In’
- Thermal stress up to 60 °C does not permanently exceed the tolerance limits
- To preserve markings, cleaning at no higher than 60 °C is recommended
Class B tolerances according to DIN EN ISO 1042
Frequently Bought Together