Orders LinkContactUsLinkComponent
Screwthread tubes, attachable, SVL® thread
Supplier:   SciLabware
Screwthread tubes, attachable, SVL® thread
Supplier:  SciLabware
Certificates
Screwthread tubes, attachable, SVL® thread
Screwthread tubes, attachable, SVL® thread
4 Options Available Below
Product Details & Documents
Made of borosilicate glas.
  • Highest resistance to thermal shock and chemical corrosion
  • Designed to use with the range of plastic screwcaps, silicone rubber rings and PTFE washers
  • Unbagged
For use by Glassblowers to construct apparatus with SVL® joints.
Product Family Options
Product Information
  • Thread
    Tube ext.Ø
    Wall thickness
    Total length
    Availability
    Price
  • SVL 15
    16 mm
    1,8 mm
    165 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 42,10 €
    42,10 €
    1 * 10 ST
       42,10 €
  • SVL 22
    22,5 mm
    1,8 mm
    165 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 210,00 €
    210,00 €
    1 * 50 ST
       210,00 €
  • SVL 30
    32 mm
    2 mm
    165 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 451,00 €
    451,00 €
    1 * 50 ST
       451,00 €
  • SVL 42
    44,1 mm
    3,2 mm
    165 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 030,00 €
    1 030,00 €
    1 * 50 ST
       1 030,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 15, ring I.Ø 10 mm
    Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 15, ring I.Ø 10 mm
    BIBB701-01
    Log in for availability
    1 * 10 ST - 69,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 22, ring I.Ø 17 mm
    Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 22, ring I.Ø 17 mm
    BIBB701-02
    Log in for availability
    1 * 10 ST - 139,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 30, ring I.Ø 23 mm
    Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 30, ring I.Ø 23 mm
    BIBB701-03
    Log in for availability
    1 * 10 ST - 142,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 42, ring I.Ø 34 mm
    Sealing rings for butt joints, silicone rubber sheathed with PTFE, SVL® joint size 42, ring I.Ø 34 mm
    BIBB701-04
    Log in for availability
    1 * 10 ST - 296,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Couplings, phenolic plastic for butt joints, SVL® joint size 42, ring I.Ø 34 mm
    Couplings, phenolic plastic for butt joints, SVL® joint size 42, ring I.Ø 34 mm
    BIBB701-14
    Log in for availability
    1 * 10 ST - 495,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 4,0 - 4,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 4,0 - 4,4 mm
    BIBB701-19
    Log in for availability
    1 * 10 ST - 113,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 4,8 - 5,2 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 4,8 - 5,2 mm
    BIBB701-20
    Log in for availability
    1 * 10 ST - 86,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 5,6 - 6,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 5,6 - 6,4 mm
    BIBB701-21
    Log in for availability
    1 * 10 ST - 90,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 5,6 - 7,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 15, for tubing O.Ø 5,6 - 7,4 mm
    BIBB701-22
    Log in for availability
    1 * 10 ST - 81,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 19,5 - 20,5 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 19,5 - 20,5 mm
    BIBB701-26
    Log in for availability
    1 * 10 ST - 189,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 21,5 - 22,5 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 21,5 - 22,5 mm
    BIBB701-27
    Log in for availability
    1 * 10 ST - 194,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 42, for tubing O.Ø 29,2 - 30,8 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 42, for tubing O.Ø 29,2 - 30,8 mm
    BIBB701-28
    Log in for availability
    1 * 10 ST - 481,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 42, for tubing O.Ø 31,2 - 32,8 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 42, for tubing O.Ø 31,2 - 32,8 mm
    BIBB701-29
    Log in for availability
    1 * 10 ST - 481,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Phenolic screw caps with hole, for SVL® joint size 15
    Phenolic screw caps with hole, for SVL® joint size 15
    BIBB701-41
    Log in for availability
    1 * 10 ST - 20,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Phenolic screw caps with hole, for SVL® joint size 22
    Phenolic screw caps with hole, for SVL® joint size 22
    212-7469
    Log in for availability
    1 * 10 ST - 23,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Phenolic screw caps with hole, for SVL® joint size 30
    Phenolic screw caps with hole, for SVL® joint size 30
    BIBB701-43
    Log in for availability
    1 * 10 ST - 29,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Phenolic screw caps with hole, for SVL® joint size 42
    Phenolic screw caps with hole, for SVL® joint size 42
    BIBB701-44
    Log in for availability
    1 * 10 ST - 84,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 11,6 - 12,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 11,6 - 12,4 mm
    BIBB701-52
    Log in for availability
    1 * 10 ST - 153,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 17,6 - 18,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 30, for tubing O.Ø 17,6 - 18,4 mm
    BIBB701-56
    Log in for availability
    1 * 10 ST - 214,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 9,6 - 10,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 9,6 - 10,4 mm
    BIBB701-51
    Log in for availability
    1 * 10 ST - 148,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 15,6 - 16,4 mm
    Sealing rings for sliding joints, silicone rubber sheathed with PTFE, SVL® joint size 22, for tubing O.Ø 15,6 - 16,4 mm
    BIBB701-25
    Log in for availability
    1 * 10 ST - 137,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Couplings, phenolic plastic for butt joints, SVL® joint size 15, ring I.Ø 10 mm
    Couplings, phenolic plastic for butt joints, SVL® joint size 15, ring I.Ø 10 mm
    BIBB701-11
    Log in for availability
    1 * 10 ST - 522,00 €