Capillary columns, Select Biodiesel
About this item
Select Biodiesel columns address the key challenge of good column lifetime when operating at very high temperatures up to 400 °C.
- Complete set of biodiesel columns for full compliance and ease-of-use
- UltiMetal technology provides high accuracy and longevity
- Designed and pre-tested for complete confidence in results
- Results are more consistent
- No column breakage
Select Biodiesel columns are offered with a pre-coupled retention gap that is leak tested prior to shipment, making life much easier for the operator. This short piece of tubing not only enhances the analytical separation but also dramatically simplifies automation when using the column with a column inlet as specified in the standard methods.
7 Options Available Below
Product Details & Documents
Select Biodiesel columns address the key challenge of good column lifetime when operating at very high temperatures up to 400 °C.
- Complete set of biodiesel columns for full compliance and ease-of-use
- UltiMetal technology provides high accuracy and longevity
- Designed and pre-tested for complete confidence in results
- Results are more consistent
- No column breakage
Select Biodiesel columns are offered with a pre-coupled retention gap that is leak tested prior to shipment, making life much easier for the operator. This short piece of tubing not only enhances the analytical separation but also dramatically simplifies automation when using the column with a column inlet as specified in the standard methods.
Product Family Options
Product Information
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice
- VARICP6530UltiMetal retention gap, methyl deactivatedUltiMetal retention gap, methyl deactivatedCapillarySelect Biodiesel0,53 mm2 mLog in for availability1 * 1 ST123,00 €
- VARICP9076For gylcerides, UltiMetal, with retention gapFor gylcerides, UltiMetal, with retention gapCapillarySelect Biodiesel0,10 µm0,32 mm10 mLog in for availability1 * 1 ST936,00 €
- VARICP9077For gylcerides, UltiMetalFor gylcerides, UltiMetalCapillarySelect Biodiesel0,10 µm0,32 mm10 mLog in for availability1 * 1 ST650,00 €
- VARICP9078For gylcerides, UltiMetal, with retention gapFor gylcerides, UltiMetal, with retention gapCapillarySelect Biodiesel0,10 µm0,32 mm15 mLog in for availability1 * 1 ST1 040,00 €
- VARICP9079For gylcerides, UltiMetalFor gylcerides, UltiMetalCapillarySelect Biodiesel0,10 µm0,32 mm15 mLog in for availability1 * 1 ST730,00 €
- VARICP9080For FAME, fused silicaFor FAME, fused silicaCapillarySelect Biodiesel0,25 µm0,32 mm30 mLog in for availability1 * 1 ST964,00 €
- VARICP9083For Methanol, fused silicaFor Methanol, fused silicaCapillarySelect Biodiesel3,00 µm0,32 mm30 mLog in for availability1 * 1 ST964,00 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.