Orders LinkContactUsLinkComponent
Protection kit
Protection kit
CPR protection kit, incl. mask, gloves and skin cleanser, packed in printed box
Catalog # 118-0024
Supplier:   ORKLA CARE
Protection kit
Catalog # 118-0024
Supplier:  ORKLA CARE
Protection kit
Protection kit
CPR protection kit, incl. mask, gloves and skin cleanser, packed in printed box
Price
11,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This protection kit is an aid in mouth-to-mouth resuscitation. It limits contact between the first aider and the person who needs resuscitation. Clear instructions are printed in colour on the plastic cloth.
Content:
1 Breathing mask with one-way valve (for mouth-to-mouth resuscitation)
2 pairs of gloves
2 Savett safety skin cleanser
Documents
Specifications
  • Description:
    CPR protection kit, incl. mask, gloves and skin cleanser, packed in printed box