HPLC columns, NUCLEOSIL® C18 Nautilus
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEOSIL® C18 Nautilus is an endcapped octadecyl modified silica phase with an embedded polar group, using for HPLC.
- Robust spherical silica gel
- Hydrophobic phase with polar properties that is suited for 100% aqueous eluents
- Separation mechanism based on hydrophobic and polar interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Specifications
- Catalog No:
- MANA720472.40
- MANA720472.46
- MANA720431.40
- MANA720430.46
- MANA720431.46
- MANA721133.30
- MANA720432.46
- MANA721649.30
- MANA720471.46
- MANA720430.40
- Carbon load:
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- 16%
- Endcapped:
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60
- L60, L60