Order Entry
Belgium
ContactUsLinkComponent
 
PIG® Sticky Steps® mat
Catalog # NEWPMAT566-BL
Supplier:  NEW PIG
undefined
PIG® Sticky Steps® mat
Catalog # NEWPMAT566-BL
Supplier:  NEW PIG

Specifications

  • Description:
    Mat
  • L×W:
    1140×610 mm
  • Packed:
    4 pads of 30 each
  • Pk:
    4

Specifications

About this item

Multi-sheet floor mat grabs grime from shoes and wheels to keep dirt out of offices, cleanrooms and other areas where clean floors are vital.

  • High-tack disposable mat pulls dirt and debris from shoes and wheels to prevent tracking into offices
  • Used sheets tear off easily and individually
  • Numbered pull-tab system shows when you're running low
  • Low-profile pack adheres to floor with easy-to-use backing
  • Each 4 mil polyethylene sheet is nonallergenic, nonodourous and nondrying
  • Walking surface: Sticky top

Weight: 6,35 kg

Thickness: 100 micron