Orders LinkContactUsLinkComponent
Spider evaporators
Spider evaporators
Supplier:   BUCHI
Spider evaporators
Supplier:  BUCHI
Certificates
Spider evaporators
Spider evaporators
7 Options Available Below
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Product Family Options
Distillation with evaporating flasks
Product Information
  • Type
    Availability
    Price
  • 531-1319
    Spider with evaporating flasks, SJ29/32, glass, 50 ml
    Spider with evaporating flasks, SJ29/32, glass, 50 ml
    For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 590,00 €
    1 590,00 €
    1 * 1 ST
       1 590,00 €
  • 531-1320
    Spider with evaporating flasks, SJ29/32, glass, 100 ml
    Spider with evaporating flasks, SJ29/32, glass, 100 ml
    For simultaneous distillation from 5×100 ml evaporating flasks with STJ 24/29
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 220,00 €
    1 220,00 €
    1 * 1 ST
       1 220,00 €
  • BUCH011574
    Spider with evaporating flasks, SJ24/40, glass, 50 ml
    Spider with evaporating flasks, SJ24/40, glass, 50 ml
    For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 300,00 €
    1 300,00 €
    1 * 1 ST
       1 300,00 €
  • BUCH011575
    Spider with evaporating flasks, SJ24/40, glass, 100 ml
    Spider with evaporating flasks, SJ24/40, glass, 100 ml
    For simultaneous distillation from 5×100 ml evaporating flasks with 24/40 NS Joint
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 470,00 €
    1 470,00 €
    1 * 1 ST
       1 470,00 €
Distillation with cylindric flasks
Product Information
  • Type
    Availability
    Price
  • 531-1321
    Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 760,00 €
    1 760,00 €
    1 * 1 ST
       1 760,00 €
  • 531-1322
    Spider with cylindric flasks, SJ29/32, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ29/32, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 12×20 ml cylindrical flasks with 29,2/32 NS Joint
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 210,00 €
    3 210,00 €
    1 * 1 ST
       3 210,00 €
  • 531-1323
    Spider with cylindric flasks, SJ29/32, glass, incl. 20 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ29/32, glass, incl. 20 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 20×20 ml cylindrical flasks with 29,2/32 NS Joint
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 350,00 €
    5 350,00 €
    1 * 1 ST
       5 350,00 €
  • BUCH011576
    Spider with cylindric flasks, SJ24/40, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ24/40, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 6×20 ml cylindrical flasks with 24/40 NS Joint
    Discontinued
  • BUCH011577
    Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint
    Discontinued
  • BUCH011578
    Spider with cylindric flasks, SJ24/40, glass, incl. 20 cylindric flasks, SJ14.5/23, 20 ml
    Spider with cylindric flasks, SJ24/40, glass, incl. 20 cylindric flasks, SJ14.5/23, 20 ml
    For simultaneous distillation from 20×20 ml cylindrical flasks with 24/40 NS Joint
    Discontinued