Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digestion system for hydroxyproline, behrotest®

Digestion system for hydroxyproline, behrotest®

Six sample digestion apparatus, complete with glassware, RH 6
Catalog # BEHRB00218426
Supplier:   BEHR
Digestion system for hydroxyproline, behrotest®
Catalog # BEHRB00218426
Supplier:  BEHR
Digestion system for hydroxyproline, behrotest®
Digestion system for hydroxyproline, behrotest®

Six sample digestion apparatus, complete with glassware, RH 6

Price
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

This six sample digestion apparatus with energy control is suitable for determination of hydroxyproline content.

Spécifications
  • Description:
    Six sample digestion apparatus, complete with glassware
  • Type:
    RH 6
  • Cat. No.:
    BEHRB00218426
  • Model:
    RH 6
  • Supply:
    230 V - 50/60 Hz
  • W×D×H:
    760×740×320 mm
  • Power:
    1960 W
  • Weight:
    19,8 kg