ContactUsLinkComponent
Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Catalog # BEHRB00218413
Supplier:  BEHR
undefined
Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Catalog # BEHRB00218413
Supplier:  BEHR
Specifications
  • Description:
    Workstation for simultaneous aqua regia digestion of up to 6 samples for heavy metals analysis, incl. microprocessor time/temp. controller
  • Environmentally Preferable:
  • Pk:
    1
  • Type:
    SMA-ARM 6
Specifications
About this item
The behrotest® SMA-ARM workstation, for simultaneous aqua regia digestion of up to six samples for heavy metals analysis, are supplied with a programmable microprocessor-controlled time/temperature controller as well as a fume manifold for drawing off the acid vapours. This results in greater convenience and functionality in routine laboratory operations.
The details of the components of the workstation are described in the table below: