Orders LinkContactUsLinkComponent
Tip boxes, dispolab®
Tip boxes, dispolab®
Supplier:   KARTELL
Tip boxes, dispolab®
Supplier:  KARTELL
Certificates
Tip boxes, dispolab®
Tip boxes, dispolab®
2 Options Available Below
Product Details & Documents
Empty boxes for use with refill tips.
  • Semitransparent boxes in PC have a horizontal sliding cover
  • Yellow boxes in PP feature a snap-on cover
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • KART9756-06
    Box for refill tips in rack, with transparent snap-on lid
    Box for refill tips in rack, with transparent snap-on lid
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 81,50 €
    81,50 €
    1 * 10 ST
       81,50 €
  • KART9756
    Box for refill filter tips in rack with transparent snap-on lid
    Box for refill filter tips in rack with transparent snap-on lid
    Grey
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 81,50 €
    81,50 €
    1 * 10 ST
       81,50 €