Orders LinkContactUsLinkComponent
Cuvettes for Amelung®
Cuvettes for Amelung®
Supplier:   KARTELL
Cuvettes for Amelung®
Supplier:  KARTELL
Certificates
Cuvettes for Amelung®
Cuvettes for Amelung®
2 Options Available Below
Product Details & Documents
Polystyrene cuvettes designed for the determination of coagulation time.
Product Family Options
Product Information
  • Volume
    Ø×H
    Availability
    Price
  • KART82631
    Amelung® cuvettes
    Amelung® cuvettes
    1 ml
    18,65/11,25×22,8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 51,50 €
    51,50 €
    1 * 1.000 ST
       51,50 €
  • KART2631
    Amelung® cuvettes with metallic mixing balls
    Amelung® cuvettes with metallic mixing balls
    1 ml
    18,65/11,25×22,8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 84,50 €
    84,50 €
    1 * 1.000 ST
       84,50 €