Order Entry
Belgium
ContactUsLinkComponent
Racks for Revco ExF, DxF and HERAfreeze® HFU B freezers
Racks for Revco ExF, DxF and HERAfreeze® HFU B freezers
Catalog # REVC920099
Supplier:  REVCO TECHNOLOGIES
undefined
Racks for Revco ExF, DxF and HERAfreeze® HFU B freezers
Catalog # REVC920099
Supplier:  REVCO TECHNOLOGIES

Specifications

  • Compartments:
    9
  • W×D×H ext.:
    140×442×240 mm
  • Pk:
    1
  • Cryobox height:
    76,2 mm

Specifications

About this item

Maximize the storage space in your ultra-low temperature freezer with these racks.