ContactUsLinkComponent
Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Catalog # 479-1248
   Low Manufacturing Impact
Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Catalog # 479-1248
   Low Manufacturing Impact
Specifications
  • Description:
    SuperSealer
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
Thermo Scientific™ SuperSealer ensures a proper seal on plates, blocks and storage tube racks.
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
  • Designed to work with Thermo Scientific Matrix SepraSeal mats and CapMats
Delivery: Includes automatic cap, tape and SepraSeal mat applicator.
Compatible sealing options
Product Information