Polystyrene standards, GPC/SEC columns calibration
About this item
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
67 Options Available Below
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certificates
Search by Lot NumberProduct Family Options
Calibration kit
Product Information
- PkAvailabilityPrice
- VARIPL2010-0102Polystyrene calibration kit, S-M2-10, 10×0,5 g
- VARIPL2010-0104Polystyrene calibration kit, S-H2-10, 10×0,5 g
- VARIPL2010-0105Polystyrene calibration kit, S-L2-10, 10×0,5 g
Individual polymer molecular weight
Product Information
- PkAvailabilityPrice
- VARIPL2012-0001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 01 g1Log in for availability1 * 1 ST201,00 €
- VARIPL2012-0005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 05 g1Log in for availability1 * 1 ST904,00 €
- VARIPL2012-1005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1, quantities 05 g1Log in for availability1 * 1 ST913,00 €
- VARIPL2012-1010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1, quantities 10 g10 gLog in for availability1 * 10 g
- VARIPL2012-3001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,09, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,09, quantities 01 g1Log in for availability1 * 1 ST164,00 €
- VARIPL2012-4001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 01 g1Log in for availability1 * 1 ST164,00 €
- VARIPL2012-4005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 05 g1Log in for availability1 * 1 ST738,00 €
- VARIPL2012-4010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,07, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2012-5001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2012-5005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2012-5010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2012-6001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2012-6005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2012-6010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2012-7001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2012-7005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2012-7010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2012-8001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2012-8005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2012-8010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2012-9001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g1Log in for availability1 * 1 ST144,00 €
- VARIPL2012-9005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2012-9010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-0001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2013-1001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g1Log in for availability1 * 1 ST142,00 €
- VARIPL2013-1005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 g1Log in for availability1 * 1 ST640,00 €
- VARIPL2013-1010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-2001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g1Log in for availability1 * 1 ST144,00 €
- VARIPL2013-2005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 g1Log in for availability1 * 1 ST646,00 €
- VARIPL2013-2010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-3001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST144,00 €
- VARIPL2013-3005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST646,00 €
- VARIPL2013-3010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-4001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST144,00 €
- VARIPL2013-4005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST646,00 €
- VARIPL2013-4010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-5001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g1Log in for availability1 * 1 ST144,00 €
- VARIPL2013-5005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 05 g1Log in for availability1 * 1 ST646,00 €
- VARIPL2013-5010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-6001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,01, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,01, quantities 01 g1Log in for availability1 * 1 ST150,00 €
- VARIPL2013-7001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 g1Log in for availability1 * 1 ST150,00 €
- VARIPL2013-7010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2013-8001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST150,00 €
- VARIPL2013-8005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST677,00 €
- VARIPL2013-9001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST150,00 €
- VARIPL2013-9005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST677,00 €
- VARIPL2013-9010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2014-0001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 01 g1Log in for availability1 * 1 ST150,00 €
- VARIPL2014-0005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 05 g1Log in for availability1 * 1 ST677,00 €
- VARIPL2014-0010Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g1Log in for availability1 * 1 ST
- VARIPL2014-1001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 g1Log in for availability1 * 1 ST166,00 €
- VARIPL2014-1005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 05 g1Log in for availability1 * 1 ST745,00 €
- VARIPL2014-2001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST166,00 €
- VARIPL2014-3001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST203,00 €
- VARIPL2014-3005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 g1Log in for availability1 * 1 ST913,00 €
- VARIPL2014-4001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,05, quantities 01 g1Log in for availability1 * 1 ST203,00 €
- VARIPL2014-5001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 g1Log in for availability1 * 1 ST203,00 €
- VARIPL2014-6001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST232,00 €
- VARIPL2014-6005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 g1Log in for availability1 * 1 ST1 040,00 €
- VARIPL2014-7001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 01 g1Log in for availability1 * 1 ST232,00 €
- VARIPL2014-7005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,04, quantities 05 g1Log in for availability1 * 1 ST1 050,00 €
- VARIPL2014-8001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 g1Log in for availability1 * 1 ST323,00 €
- VARIPL2014-8005Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 05 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 05 g1Log in for availability1 * 1 ST1 460,00 €
- VARIPL2014-9001Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 gPolystyrene, individual polymer molecular weight, nominal Mw/Mn 1,06, quantities 01 g1Log in for availability1 * 1 ST320,00 €