Distillation apparatus, Behrotest® WE 5 and associated glassware
Supplier: BEHR
Certificates
About this item
The behrotest® WE 5 Five-in-a-line multiple steam generator provides the simultaneously rapid and gentle distillation of up to five samples. The discharge of steam into the samples guarantees a particularly uniform distillation and thus a high reproducibility of the recovery rates over the entire range. The complete set includes:
- Temperature and time controller
- Heating block with five cavities
- Thermal insulation cover
- Stand for DET 5 glassware
- Coil cooler
For the determination of ammonium-nitrogen as defined in DIN 38406-E5 and DIN 38409-H16-2 phenol index procedure.
Frequently Bought Together