Order Entry
Belgium
ContactUsLinkComponent
Dispensing closures for bottles
Dispensing closures for bottles
  MURRP516/15
 :  MURRAY S
Dispensing closures for bottles
  MURRP516/15
 :  MURRAY S

 

  • Thread:
    22 mm R6
  • Colour:
    White
  • Description:
    with flip-up spout, LDPE, aperture 2 mm
  • Pk:
    100
  • For bottles:
    60/125/175 and 250 ml