Orders LinkContactUsLinkComponent
Jars, tall
Jars, tall
Supplier:   MURRAY S
Jars, tall
Supplier:  MURRAY S
Certificates
Jars, tall
10 Options Available Below
Product Details & Documents
Clear or amber glass, without cap.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Amber coloured to protect light-sensitive materials
Perfect for sample collection, storing and transporting of powders, granulates, pastes or liquids.
Product Family Options
Clear glass
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 30 ml
    39 mm
    59 mm
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 88,90 €
    88,90 €
    1 * 80 ST
       88,90 €
  • 500 ml
    83 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 33 ST
    1 * 33 ST - 123,00 €
    123,00 €
    1 * 33 ST
       123,00 €
  • 60 ml
    45 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 86,80 €
    86,80 €
    1 * 72 ST
       86,80 €
  • 114 ml
    54 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 113,00 €
    113,00 €
    1 * 80 ST
       113,00 €
  • 227 ml
    64 mm
    119 mm
    Log in for availability
    Log in for availability
    1 * 42 ST
    1 * 42 ST - 64,90 €
    64,90 €
    1 * 42 ST
       64,90 €
Amber glass
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 30 ml
    39 mm
    59 mm
    Log in for availability
    Log in for availability
    1 * 127 ST
    1 * 127 ST - 96,20 €
    96,20 €
    1 * 127 ST
       96,20 €
  • 60 ml
    45 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 70,70 €
    70,70 €
    1 * 72 ST
       70,70 €
  • 114 ml
    54 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 68 ST
    1 * 68 ST - 80,20 €
    80,20 €
    1 * 68 ST
       80,20 €
  • 227 ml
    64 mm
    59 mm
    Log in for availability
    Log in for availability
    1 * 42 ST
    1 * 42 ST - 54,50 €
    54,50 €
    1 * 42 ST
       54,50 €
  • 500 ml
    83 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 28 ST
    1 * 28 ST - 102,00 €
    102,00 €
    1 * 28 ST
       102,00 €