Orders LinkContactUsLinkComponent
Preparative columns, PLgel™
Preparative columns, PLgel™
Supplier:   VARIAN
Preparative columns, PLgel™
Supplier:  VARIAN
Certificates
Preparative columns, PLgel™
10 Options Available Below
Product Details & Documents
These preparative columns is ideal to fractionate polymers, isolate components in a polymer formulation or simplify mixtures of relatively small molecules in complex matrices.
  • Excellent column efficiency provides optimum resolution
  • High loading can isolate milli grams amounts for further study
  • Over 10 times scale up permits efficient quantification
It is packed with the same rigid, high performance media as the analytical columns so that mixtures of materials are easily separated on the basis of size, preferably in a low boiling organic solvent. They are then collected as a series of discrete fractions and isolated by simple evaporation of the solvent.
Product Family Options
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • VARIPL1210-6115
    PLgel™, MW ranges upto 2000
    PLgel™, MW ranges upto 2000
    Preparative
    50 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 780,00 €
    4 780,00 €
    1 * 1 ST
       4 780,00 €
  • VARIPL1210-6120
    PLgel™, MW ranges upto 4000
    PLgel™, MW ranges upto 4000
    Preparative
    100 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 780,00 €
    4 780,00 €
    1 * 1 ST
       4 780,00 €
  • VARIPL1210-6125
    PLgel™, MW ranges 500 - 30000
    PLgel™, MW ranges 500 - 30000
    Preparative
    500 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 880,00 €
    4 880,00 €
    1 * 1 ST
       4 880,00 €
  • VARIPL1210-6130
    PLgel™, MW ranges 500 - 60000
    PLgel™, MW ranges 500 - 60000
    Preparative
    1000 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 970,00 €
    4 970,00 €
    1 * 1 ST
       4 970,00 €
  • VARIPL1210-6140
    PLgel™, MW ranges 10000 - 600000
    PLgel™, MW ranges 10000 - 600000
    Preparative
    10000 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 970,00 €
    4 970,00 €
    1 * 1 ST
       4 970,00 €
  • VARIPL1210-6150
    PLgel™, MW ranges 60000 - 2000000
    PLgel™, MW ranges 60000 - 2000000
    Preparative
    100000 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 970,00 €
    4 970,00 €
    1 * 1 ST
       4 970,00 €
  • VARIPL1210-6160
    PLgel™, MW ranges 600000 - 10000000
    PLgel™, MW ranges 600000 - 10000000
    Preparative
    1000000 Å
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 880,00 €
    4 880,00 €
    1 * 1 ST
       4 880,00 €
  • VARIPL1210-6104
    PLgel™, Mixed-D, MW ranges 200 - 400000
    PLgel™, Mixed-D, MW ranges 200 - 400000
    Preparative
    -
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • VARIPL1210-6100
    PLgel™, Mixed-B, MW ranges 500 - 10000000
    PLgel™, Mixed-B, MW ranges 500 - 10000000
    Preparative
    -
    10 µm
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 580,00 €
    5 580,00 €
    1 * 1 ST
       5 580,00 €
  • VARIPL1210-1120
    PLgel™, Prep guard
    PLgel™, Prep guard
    Guard column
    -
    - µm
    25,00 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 530,00 €
    1 530,00 €
    1 * 1 ST
       1 530,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.