Orders LinkContactUsLinkComponent
Dressing jars
Dressing jars
Supplier:   MURRAY S
Dressing jars
Supplier:  MURRAY S
Certificates
17 Options Available Below
Product Details & Documents
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered separately
Product Family Options
Product Information
  • Ext.Ø
    Height
    Availability
    Price
  • 100 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,70 €
    37,70 €
    1 * 1 ST
       37,70 €
  • 125 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,60 €
    43,60 €
    1 * 1 ST
       43,60 €
  • 100 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,60 €
    43,60 €
    1 * 1 ST
       43,60 €
  • 150 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • 100 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,60 €
    43,60 €
    1 * 1 ST
       43,60 €
  • 150 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,10 €
    72,10 €
    1 * 1 ST
       72,10 €
  • 200 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,10 €
    94,10 €
    1 * 1 ST
       94,10 €
  • 150 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,50 €
    84,50 €
    1 * 1 ST
       84,50 €
  • 200 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €
  • 200 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
Lids
Product Information
  • Ext.Ø
    Height
    Availability
    Price
  • MURRG501/01
    Lids, moulded glass, circular, for jars of O.Ø 100 mm
    Lids, moulded glass, circular, for jars of O.Ø 100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,50 €
    33,50 €
    1 * 1 ST
       33,50 €
  • MURRG501/02
    Lids, moulded glass, circular, for jars of O.Ø 125 mm
    Lids, moulded glass, circular, for jars of O.Ø 125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,00 €
    28,00 €
    1 * 1 ST
       28,00 €
  • MURRG501/03
    Lids, moulded glass, circular, for jars of O.Ø 150 mm
    Lids, moulded glass, circular, for jars of O.Ø 150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,70 €
    30,70 €
    1 * 1 ST
       30,70 €
  • MURRG502/01
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 100 mm
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,60 €
    31,60 €
    1 * 1 ST
       31,60 €
  • MURRG502/02
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 125 mm
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,70 €
    35,70 €
    1 * 1 ST
       35,70 €
  • MURRG502/03
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 150 mm
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,30 €
    39,30 €
    1 * 1 ST
       39,30 €
  • MURRG502/04
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,60 €
    45,60 €
    1 * 1 ST
       45,60 €