Orders LinkContactUsLinkComponent
Dissection sets
Supplier:   MURRAY S
Dissection sets
Supplier:  MURRAY S
Certificates
Dissection sets
2 Options Available Below
Product Details & Documents
These high quality dissecting instruments are designed for everyday use. They are subject to stringent manufacturing quality procedures, and therefore suitable for surgical applications.
The Basic Set includes:
Two scalpels
Two needles
One seeker
One blunt forceps
One sharp forceps
One fine scissors
One blunt scissors
The Advanced Set includes:
Three scalpels
Two needles
One seeker
One section lifter
One blunt forceps
One sharp forceps
One sharp curved forceps
One fine scissors
One blunt scissors
One section razor
Product Family Options
Product Information
  • Availability
    Price
  • 631-1092
    Dissection kit, Basic Set in PVC roll, 9 pcs.
    Dissection kit, Basic Set in PVC roll, 9 pcs.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,30 €
    35,30 €
    1 * 1 ST
       35,30 €
  • 631-1093
    Dissection kit, Advanced Set in PVC roll, 13 pcs.
    Dissection kit, Advanced Set in PVC roll, 13 pcs.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,10 €
    65,10 €
    1 * 1 ST
       65,10 €