Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps, SAMCO

Forceps, SAMCO

Pointed, 125 mm
Catalog # MURRL760/01
Supplier:   MURRAY S
Forceps, SAMCO
Catalog # MURRL760/01
Supplier:  MURRAY S
Forceps, SAMCO
Forceps, SAMCO

Pointed, 125 mm

Price
90,20 €
Log in for availability
1 * 100 ST
Product Details & Documents

Glass filled nylon, white.

  • Spring forceps for dissecting applications
  • Autoclavable to 125 °C
Specifications
  • Type:
    Pointed
  • Length:
    125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.