Orders LinkContactUsLinkComponent
Stoppers for sample bottles, BEHROTEST®
Supplier:   BEHR
Stoppers for sample bottles, BEHROTEST®
Supplier:  BEHR
Certificates
Stoppers for sample bottles, BEHROTEST®
Stoppers for sample bottles, BEHROTEST®
2 Options Available Below
Product Details & Documents
These glass stoppers can be used instead of DIN/DEV diagonally cut stoppers. They are easy to use due to the lack of the easily broken edges.
  • No formation of bubbles below the stopper tip on inserting the stopper into the liquid sample
  • Less breakage or cracking risk than hollow stoppers
Product Family Options
Product Information
  • NS
    Availability
    Price
  • 29/32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,00 €
    33,00 €
    1 * 1 ST
       33,00 €
  • 24/29
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,50 €
    18,50 €
    1 * 1 ST
       18,50 €