ContactUsLinkComponent
Accessories for 2D barcode reader, VisionMate® High Speed
Accessories for 2D barcode reader, VisionMate® High Speed
Catalog # MATRAB-1860
undefined
Accessories for 2D barcode reader, VisionMate® High Speed
Catalog # MATRAB-1860
Specifications
  • Description:
    1D reader adapter, for reading and integrating codes on the sides of racks into file names
  • W×D×H:
    57×101×133 mm
  • Interfaces:
    USB
  • Weight:
    500 g
  • Pk:
    1